missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Kartogenin

Product Code. 17120092
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17120092 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17120092 Supplier STEMCELL Technologies Supplier No. 72572

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

RUNX1 transcriptional activator; Binds filamin A

  • Kartogenin induces chondrogenesis by binding the actin-binding protein, filamin A, which disrupts its interaction with the transcription factor core-binding factor β subunit (CBFβ)
  • When dissociated from filamin A, CBFβ translocates to the nucleus and forms a transcriptional complex with the runt-related transcription factor RUNX1, which enables chondrocyte differentiation (Johnson et al.)
  • Promotes differentiation of human bone marrow mesenchymal stem cells into chondrocytes (Johnson et al.; Zhang et al.)
  • Promotes cartilage formation/repair in mouse models of osteoarthritis or when injected into mouse tendon-bone junctions (Johnson et al.; Zhang et al.)
  • Promotes type-I collagen synthesis in human dermal fibroblasts in vitro and in the dermis of mice through activation of the SMAD4/SMAD5 pathway (Wang et al.)

Specifications

CAS 4727-31-5
Quantity 5 mg
Molecular Formula C20H15NO3
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.