missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Kartogenin

Product Code. 16554434 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16554434 250 mg 250mg
16544434 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16554434 Supplier Thermo Scientific Chemicals Supplier No. 464662500
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4727-31-5
Molecular Formula C20H15NO3
Molecular Weight (g/mol) 317.34
InChI Key SLUINPGXGFUMLL-UHFFFAOYSA-N
IUPAC Name 2-({[1,1'-biphenyl]-4-yl}carbamoyl)benzoic acid
SMILES OC(=O)C1=CC=CC=C1C(=O)NC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point >280.0°C
Quantity 250 mg
Solubility Information Solubility aq. soln.: .. Other solubilities: slightly soluble in dmso and methanol
Formula Weight 317.34
Chemical Name or Material Kartogenin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.