missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KARS Recombinant Rabbit Monoclonal Antibody (2V2B6)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538171
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, 293T, C6, Mouse testis, Rat testis Immunogen sequence: TELNDPMRQR QLFEEQAKAK AAGDDEAMFI DENFCTALEY GLPPTAGWGM GIDRVAMFLT DSNNIKEVLL FPAMKPEDKK ENVATTDTLE STTVGTSV.
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Lysyl-tRNA synthetase is a homodimer localized to the cytoplasm which belongs to the class II family of tRNA synthetases. It has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| KARS | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15046, Q5XIM7, Q99MN1 | |
| KARS1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 500-597 of human LysRS/KARS (Q15046). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 2V2B6 | |
| Unconjugated | |
| KARS1 | |
| AA589550; AL024334; AL033315; AL033367; CMTRIB; D8Ertd698e; D8Wsu108e; DFNB89; EC 6.1.1.6; KARS; Kars1; KARS2; KIAA0070; KRS; lysine tRNA ligase; lysine--tRNA ligase; LysRS; lysyl-tRNA synthetase; mKIAA0070 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 292028, 3735, 85305 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction