missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KARS Recombinant Rabbit Monoclonal Antibody (23GB6410), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | KARS |
|---|---|
| Clone | 23GB6410 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087507
![]() |
Invitrogen™
MA553042 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088814
|
Invitrogen™
MA553043 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KARS Recombinant Monoclonal antibody specifically detects KARS in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| KARS | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q15046, Q5XIM7, Q99MN1 | |
| 292028, 3735, 85305 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6410 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AA589550; AL024334; AL033315; AL033367; CMTRIB; D8Ertd698e; D8Wsu108e; DFNB89; EC 6.1.1.6; KARS; Kars1; KARS2; KIAA0070; KRS; lysine tRNA ligase; lysine--tRNA ligase; LysRS; lysyl-tRNA synthetase; mKIAA0070 | |
| A synthesized peptide derived from human LysRS (550-597AA). | |
| Primary | |
| -20°C | |
| KARS1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title