missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Fischer Standard: 1.0 wt% H2O in 10W30 Motor Oil, VHG™
Description
Karl Fischer Standard: 1.0 wt% H2O in 10W30 Motor Oil
Specifications
Specifications
| Chemical Name or Material | Nominal Water |
| Methods | Coulometric Titration |
| Analyte or Component Names | Nominal Water |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| IUPAC Name | Nominal Water |
| Solvent or Matrix | 10W30 Motor Oil |
| Quantity | 100 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?