missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Fischer Aqualine™ Electrolyte AG-H, for Karl Fischer Titration By Coulometry, Fisher Chemical
Shop All Fisher Chemical ProductsSpecifications
Specifications
| Physical Form | Liquid |
| Quantity | 500 mL |
| MDL Number | 81734 |
| Packaging | Amber glass bottle |
Safety and Handling
Product Identifier
- Aqualine Electrolyte AG-H
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Methyl alcohol, Amylalcohol, 2-Amino-2-methyl-1-propanol, 2, 4, 6-Collidine, Sulfur dioxide, Iodine
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?