missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KappaB ras2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100869
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NKIRAS2.
Atypical Ras-like protein that acts as a potent regulator of NF-kappa-B activity by preventing the degradation of NF-kappa-B inhibitor beta (NFKBIB) by most signals, explaining why NFKBIB is more resistant to degradation. May act by blocking phosphorylation of NFKBIB and nuclear localization of p65/RELA NF-kappa-B subunit. It is unclear whether it acts as a GTPase. Both GTP- and GDP-bound forms block phosphorylation of NFKBIB.
Specifications
| KappaB ras2 | |
| Polyclonal | |
| Unconjugated | |
| NKIRAS2 | |
| 2410003M04Rik; 4930527H08Rik; D630018G21Rik; I-kappa-B-interacting Ras-like protein 2; Kappa B-Ras protein 2; kappaB-Ras2; KBRAS2; NF-kappa-B inhibitor-interacting Ras-like protein 2; NFKB inhibitor interacting Ras like 2; NFKB inhibitor interacting Ras-like 2; NFKB inhibitor interacting Ras-like protein 2; NKIRAS2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 28511, 71966 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9CR56, Q9NYR9 | |
| NKIRAS2 | |
| A synthesized peptide derived from human NKIRAS2(Accession Q9NYR9), corresponding to amino acid residues A152-G191. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction