missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KappaB ras1 Monoclonal Antibody (184C278)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA141180
This item is not returnable.
View return policy
Description
Suggested positive control: 293 whole cell lysate. Immunogen sequence: RVELLKKEID KFKDKKEV.
KappaB ras-1 (kappa-B-ras-1) and kappaB-ras-2 are two small proteins that similar to Ras-like small GTPases that associate with Ikappa-B (Ikappa-B), an inhibitor of the transcription factor NF-kappa-B. Ikappa-B exists in two homologous forms, Ikappa-B-alpha and Ikappa-B-beta, although Ikappa-B-beta contains a unique 47-amino acid region within its ankyrin domain. While inactive Ikappa-B-alpha-NF-kappa-B complexes can shuttle in and out of the nucleus, Ikappa-B-beta-NF-kappa-B complexes are retained exclusively in the cytoplasm. It is suggested that kappaB-ras proteins preferentially bind to the Ikappa-B-beta form through this unique insert within the ankyrin region, thus modulating the cellular location of Ikappa-B-beta and regulating the rate of degradation of Ikappa-B-beta.
Specifications
| KappaB ras1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.2% gelatin and 0.05% sodium azide | |
| Q8CEC5, Q9NYS0 | |
| NKIRAS1, nkiras1.S | |
| Synthetic peptide containing amino acid residues 96-113 of human kB-Ras1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Bovine, Pig, Horse, Frog | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| 184C278 | |
| Unconjugated | |
| NKIRAS1 | |
| 2400004O09Rik; C15orf1; I1PP2A; I-kappa-B-interacting Ras-like protein 1; kappa B-ras 1; kappa B-Ras protein 1; kappaB-Ras1; KBRAS1; LANP; MAPM; MGC119787; MGC150373; NF-kappa-B inhibitor-interacting Ras-like protein 1; NFKB inhibitor interacting Ras like 1; NFKB inhibitor interacting Ras-like 1; NFKB inhibitor interacting Ras-like 1 S homeolog; NFKB inhibitor interacting Ras-like protein 1; Nkiras1; nkiras1.S; PHAP1; PHAPI; PP32; XELAEV_18033117mg | |
| Mouse | |
| Protein G | |
| RUO | |
| 100036787, 100051752, 100620195, 28512, 305751, 477047, 522875, 69721 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction