missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Kappa Light Chain Recombinant Rabbit Monoclonal Antibody (K01_2Q22)

Product Code. 30830329
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830329 50 μL
30827810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830329 Supplier Invitrogen™ Supplier No. MA565712
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Kappa Light Chain Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

Immunoglobulin classes share the same basic four polypeptide chain structure of two heavy chains (five types) and two light chains (kappa, lambda; both having a molecular weight of 22.5 kDa). Kappa and lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region. The ratio of kappa to lambda is 70:30. Antibodies to the kappa light chain are reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kappa Light Chain
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2Q22
Concentration 0.70 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene IGK
Gene Accession No. P01601, P01602, P01834, P0DOX7
Gene Alias D4Mit8; HCAK1; ig kappa chain c region; Igk; Igk@; Igkc; Igk-C; IGKCD; IGKV; IGKV15; IGKV1-5; Immunoglobulin K; immunoglobulin kappa chain complex; immunoglobulin kappa constant; immunoglobulin kappa light chain; immunoglobulin kappa locus; immunoglobulin kappa variable 1-5; kappa; Kappa LC; Km; L12; L12a; LOC500183; similar to NGF-binding Ig light chain; V1
Gene Symbols IGK, IGKC, IGKV1-5, IGKV1D-16
Host Species Rabbit
Immunogen A synthesized peptide derived from human Human Kappa Chain.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28299, 28901, 3514, 50802
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.