missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappa Light Chain Monoclonal Antibody (TB28-2), Super Bright™ 600, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£139.00 - £354.00
Specifications
| Antigen | Kappa Light Chain |
|---|---|
| Clone | TB28-2 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16331544
|
Affymetrix eBioscience
63-9970-41 |
25 Tests | |||||||
|
16341544
|
Invitrogen™
63-9970-42 |
100 Tests |
£354.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mouse Monoclonal Antibody
Immunoglobulin classes share the same basic four polypeptide chain structure of two heavy chains (five types) and two light chains (kappa, lambda; both having a molecular weight of 22.5 kDa). Kappa and lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region. The ratio of kappa to lambda is 70:30. Antibodies to the kappa light chain are reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Specifications
| Kappa Light Chain | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! |
| TB28-2 | |
| Flow Cytometry | |
| Super Bright 600 | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title