missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Kappa Light Chain Monoclonal antibody specifically detects Kappa Light Chain in Human, Rat (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Kappa Light Chain |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SPM508 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.5-2.0 ug/million cells, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 ug/mL |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Accession No. | P01601, P01834 |
| Gene Alias | HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?