missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappa Light Chain Antibody (MEM-09), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-331PECY55
This item is not returnable.
View return policy
Description
Kappa Light Chain Monoclonal specifically detects Kappa Light Chain in Human samples. It is validated for Flow Cytometry.
Specifications
| Kappa Light Chain | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| IGKC | |
| Crude thymus membrane fraction. | |
| 0.1 mL | |
| Primary | |
| The antibody MEM-09 reacts with both secreted and B cell-surface human immunoglobulin, specifically reacts with kappa light chains (22.5 kDa). Material immunoprecipitated from human serum with the antibody MEM-09 consists of IgG and traces of IgM. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| MEM-09 | |
| Flow Cytometry | |
| HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, Immunology | |
| 3514.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction