missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappa Light Chain Antibody (Kap-56), Allophycocyanin , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11599APC
This item is not returnable.
View return policy
Description
Kappa Light Chain Monoclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for Flow Cytometry
Specifications
| Kappa Light Chain | |
| Monoclonal | |
| APC | |
| HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
| Human lymphocytes stimulated | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3514 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| Kap-56 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction