missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappa Light Chain Antibody (H16-E), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP1-30140-0.1ml
This item is not returnable.
View return policy
Description
Kappa Light Chain Monoclonal specifically detects Kappa Light Chain in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Kappa Light Chain | |
| Monoclonal | |
| Unconjugated | |
| 20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.5% Sodium Azide | |
| HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody is specific to the C terminus of Human IGKC | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| H16-E | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:500 | |
| P01834 | |
| IGKC | |
| Peptide derived from C-terminal sequence of IGKC | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3514 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction