missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappa Light Chain Antibody (4G7cc), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-7084H
This item is not returnable.
View return policy
Description
Kappa Light Chain Monoclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Kappa Light Chain | |
| Monoclonal | |
| HRP | |
| HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with a mixture of light chains of human immunoglobulin. | |
| 0.2 mL | |
| Adaptive Immunity, Immunology | |
| 3514 | |
| Store at 4°C in the dark. | |
| IgG2a |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 4G7cc | |
| PBS | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction