missing translation for 'onlineSavingsMsg'
Learn More

Kappa Light Chain Ab-2 Rabbit Polyclonal Antibody, Epredia™

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Kappa Light Chain Ab-2, Rabbit Polyclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

£122.40 - £384.61

Specifications

Antigen Kappa Light Chain Ab-2
Clone Polyclonal
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12613227
View Documents
Epredia
RB-333-A0
100 μL Concentrated This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12623227
View Documents
Epredia
RB-333-A1
500 μL Concentrated
ONLINE EXCLUSIVE PRICE
£249.00
£233.65 / 500µL
Save £15.35
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12324173
View Documents
Epredia
RB-333-A
1 mL Concentrated
ONLINE EXCLUSIVE PRICE
£419.00
£384.61 / 1mL
Save £34.39
8% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12653227
View Documents
Epredia
RB-333-R7
7 mL Ready-to-Use This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12663227
View Documents
Epredia
RB-333-RQ
12 mL Ready-to-Use This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Antibody to the κ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Baboon, Monkey and Rat. Does not react with dog. Others not known.

Immunogen: Kappa light chains purified from human Bence Jones proteins

Molecular Weight: 25kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Kappa Light Chain Ab-2
Immunohistochemistry (Paraffin)
Unconjugated
IVD
kappa light chains purified from human Bence Jones proteins
Polyclonal
Polyclonal
Rabbit
Hematopoietic
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.