missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KAP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554878
This item is not returnable.
View return policy
Description
Immunogen sequence: ELLNAQQEDD EFVCQIIYVF YQMVFHQATR DVIIKETQAP AYLIDLMHDK NNEIRKVCDN TLDIIAEYDE EWAKK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The small G protein GDP dissociation stimulator (smg GDS) is a regulator protein having two activities on a group of small G proteins including the Rho and Rap1 family members and Ki-Ras; one is to stimulate their GDP/GTP exchange reactions, and the other is to inhibit their interactions with membranes. The protein encoded by this gene contains 9 'Armadillo' repeats and interacts with the smg GDS protein through these repeats. This protein, which is highly concentrated around the endoplasmic reticulum, is phosphorylated by v-src, and this phosphorylation reduces the affinity of the protein for smg GDS. It is thought that this protein serves as a linker between human chromosome-associated polypeptide (HCAP) and KIF3A/B, a kinesin superfamily protein in the nucleus, and that it plays a role in the interaction of chromosomes with an ATPase motor protein. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| KAP3 | |
| Polyclonal | |
| Unconjugated | |
| KIFAP3 | |
| dJ190I16.1; FLA3; KAP-1; KAP3; KAP-3; KIF3AP; Kifap3; kinesin associated protein 3; kinesin-associated protein 3; small G protein GDP dissociation stimulator; SMAP; Smg GDS-associated protein; Smg-GDS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22920 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92845 | |
| KIFAP3 | |
| Recombinant protein corresponding to Human KAP3. Recombinant protein control fragment (Product #RP-94021). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction