missing translation for 'onlineSavingsMsg'
Learn More

KAP1 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18429877 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429877 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429877 Supplier Novus Biologicals Supplier No. NBP232072FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KAP1 Polyclonal antibody specifically detects KAP1 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KAP1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias E3 SUMO-protein ligase TRIM28, EC 6.3.2.-, FLJ29029, KAP-1, KAP1KRAB-associated protein 1, KRIP-1, Nuclear corepressor KAP-1, RNF96KRAB-interacting protein 1, TF1B, TIF1-beta, TIF1BRING finger protein 96, transcription intermediary factor 1-beta, transcriptional intermediary factor 1-beta, tripartite motif containing 28, tripartite motif-containing 28, Tripartite motif-containing protein 28
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 550 to 600 of human Tripartite Motif-Containing 28 using the numbering given in entry NP_035718.2 (GeneID 21849).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 10155
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.