missing translation for 'onlineSavingsMsg'
Learn More

Kaolin, TRC

Product Code. 30403007
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403007 100 g 100g
30400098 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30403007 Supplier Toronto Research Chemicals Supplier No. TRCK139560100G
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Kaolin
CAS 1332-58-7
Molecular Formula Al2O7Si2.2H2O
Quantity 100 g
Synonym Glomax, China clay, Hydrated aluminum silicate, Kaolin clay slurry
InChI Formula InChI=1S/2Al.O5Si2.2H2O.2O/c;;1-6(2)5-7(3)4;;;;/h;;;2*1H2;;/q2*+1;-2;;;;
SMILES O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O
IUPAC Name oxo-oxoalumanyloxy-[oxo(oxoalumanyloxy)silyl]oxy-silane;dihydrate
Molecular Weight (g/mol) 258.16
Formula Weight 257.9025
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.