missing translation for 'onlineSavingsMsg'
Learn More

Kaolin, Technical, Fisher Chemical

Product Code. 11453703 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
3 kg
Packaging:
Plastic keg
Unit Size:
3kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11453703 3 kg Plastic keg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453703 Supplier Fisher Chemical Supplier No. K/0045/63

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 1760°C
CAS 1332-58-7
Color Beige
Physical Form Powder/Solid
Quantity 3 kg
Molecular Formula Al2H4O9Si2
MDL Number MFCD00062311
Synonym kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss
InChI Key NLYAJNPCOHFWQQ-UHFFFAOYSA-N
SMILES O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O
IUPAC Name oxo-oxoalumanyloxy-[oxo(oxoalumanyloxy)silyl]oxysilane;dihydrate
Molecular Weight (g/mol) 258.16
PubChem CID 56841936
Formula Weight 258.2g/mol
Packaging Plastic keg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.