missing translation for 'onlineSavingsMsg'
Learn More

Kaolin hydrated aluminum silicate, MP Biomedicals™

Product Code. 11493332 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493332 500 g 500g
11403342 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493332 Supplier MP Biomedicals Supplier No. 0215514490
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1332-58-7
Molecular Formula Al2H4O9Si2
Molecular Weight (g/mol) 258.16
MDL Number MFCD00062311
InChI Key NLYAJNPCOHFWQQ-UHFFFAOYSA-N
Synonym kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss
PubChem CID 56841936
IUPAC Name dioxo-2,4,6-trioxa-3,5-disila-1,7-dialuminaheptane-3,5-dione dihydrate
SMILES O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O

Specifications

Color White
Physical Form Powder
Quantity 500 g
Density 2.58g/mL
Chemical Name or Material Kaolin hydrated aluminum silicate
compliance-icons compliance-icons
Product Identifier
  • Kaolin Hydrated Aluminum Silicate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: kaolin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.