missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KANSL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570090
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
This gene encodes a nuclear protein that is a subunit of two protein complexes involved with histone acetylation, the MLL1 complex and the NSL1 complex. The corresponding protein in Drosophila interacts with K(lysine) acetyltransferase 8, which is also a subunit of both the MLL1 and NSL1 complexes.
Specifications
| KANSL1 | |
| Polyclonal | |
| Unconjugated | |
| KANSL1 | |
| 1700081L11Rik; 9430041J06Rik; CENP-36; centromere protein 36; centromere protein 36 antibody; DKFZp686P06109 antibody; DKFZp727C091 antibody; hMSL1v1; Kansl1; KAT8 regulatory NSL complex subunit 1; KAT8 regulatory NSL complex subunit 1 antibody; KDVS; KIAA1267; KIAA1267 antibody; male-specific lethal 1 homolog; MGC102843 antibody; mKIAA1267; MLL1/MLL complex subunit KANSL1; MLL1/MLL complex subunit male-specific lethal 1 homolog antibody; MSL1 homolog 1; MSL1V1; MSL1v1 antibody; non-specific lethal 1 homolog; NSL complex protein NSL1; NSL1; RGD1311429 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 284058 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7Z3B3 | |
| KANSL1 | |
| synthetic peptide directed towards the middle region of human KANSL1 (aa 128-177). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction