missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KANK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20546-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to KANK2.
This gene encodes a member of the KN motif and ankyrin repeat domains (KANK) family of proteins, which play a role in cytoskeletal formation by regulating actin polymerization. The encoded protein functions in the sequestration of steroid receptor coactivators and possibly other proteins. Mutations in this gene are associated with impaired kidney podocyte function and nephrotic syndrome, and keratoderma and woolly hair.Specifications
| KANK2 | |
| Polyclonal | |
| Unconjugated | |
| KANK2 | |
| ANKRD25, KANK2, KIAA1518, MXRA3, SIP, SRC 1 interacting protein, SRC1 interacting protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100361376, 235041, 25959 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q63ZY3, Q8BX02 | |
| Kank2 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction