Learn More
Invitrogen™ KANK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116620
Description
Positive Samples: Mouse skeletal muscle, Mouse heart, Mouse kidney Immunogen sequence: MAQVLHVPAP FPGTPGPASP PAFPAKDPDP PYSVETPYGY RLDLDFLKYV DDIEKGHTLR RVAVQRRPRL SSLPRGPGSW WTSTESLCSN ASGDSRHSAY SYCGRGFYPQ YGALETRGGF NPRVERTLLD ARRRLEDQAA TPTGLGSLTP SAAGSTASLV GVGLPPPTPR SSGLSTPVPP SAGHLAHVRE QMAGALRKLR QLEEQVKLIP VLQVKLSVLQ EEKRQLTVQL KSQKFLGHPT AGRGRSELCL DLPDPPEDPV ALETRSVGTW VRERDLGMPD GEAALAAKVA VLETQLKKAL.
This gene encodes a member of the KN motif and ankyrin repeat domains (KANK) family of proteins, which play a role in cytoskeletal formation by regulating actin polymerization. The encoded protein functions in the sequestration of steroid receptor coactivators and possibly other proteins. Mutations in this gene are associated with impaired kidney podocyte function and nephrotic syndrome, and keratoderma and woolly hair.
Specifications
| KANK2 | |
| Polyclonal | |
| Unconjugated | |
| KANK2 | |
| AI504612; ANKRD25; ankyrin repeat domain 25; ankyrin repeat domain-containing protein 25; BC010245; Kank2; KIAA1518; kidney ankyrin repeat-containing protein 2; KN motif and ankyrin repeat domain-containing protein 2; KN motif and ankyrin repeat domains 2; matrix-remodeling-associated protein 3; MXRA3; PPKWH; RGD1563532; SIP; SRC-1-interacting protein; SRC1-interacting protein; SRC-interacting protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100361376, 235041, 25959 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.33 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q63ZY3, Q8BX02 | |
| KANK2 | |
| Recombinant Protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.