missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KANK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£196.00 - £470.00
Specifications
| Antigen | KANK2 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18453841
|
Novus Biologicals
NBP1-89080-25ul |
25 μL |
£196.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18261308
|
Novus Biologicals
NBP1-89080 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KANK2 Polyclonal specifically detects KANK2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| KANK2 | |
| Polyclonal | |
| Rabbit | |
| Human, Rat | |
| ANKRD25, ankyrin repeat domain 25, Ankyrin repeat domain-containing protein 25, DKFZp434N161, FLJ20004, KIAA1518SRC1-interacting protein, kidney ankyrin repeat-containing protein 2, KN motif and ankyrin repeat domain-containing protein 2, KN motif and ankyrin repeat domains 2, Matrix-remodeling-associated protein 3, matrix-remodelling associated 3, MGC119707, MXRA3, SIP, SRC-1 interacting protein, SRC-1-interacting protein | |
| KANK2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 25959 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SQHIPTAEGASGSNTEEEIRMELSPDLISACLALEKYLDNPNALTERELKVAYTTVLQEWLRLACRSDAHPELVRRHLVTFRAMSARLLDYVVNIADSNGNTALHYSVSHANFPVVQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title