missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KANK2 Polyclonal antibody specifically detects KANK2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | KANK2 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ANKRD25, ankyrin repeat domain 25, Ankyrin repeat domain-containing protein 25, DKFZp434N161, FLJ20004, KIAA1518SRC1-interacting protein, kidney ankyrin repeat-containing protein 2, KN motif and ankyrin repeat domain-containing protein 2, KN motif and ankyrin repeat domains 2, Matrix-remodeling-associated protein 3, matrix-remodelling associated 3, MGC119707, MXRA3, SIP, SRC-1 interacting protein, SRC-1-interacting protein |
| Immunogen | KANK2 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human KANK2 . The immunogen is located within the last 50 amino acids of KANK2. Amino Acid Squence: CAPMSDDESPTSSSAEE |
| Quantity | 0.1 mL |
| Primary or Secondary | Primary |
| Content And Storage | Store at 4C in the dark. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?