missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin Sulfate, Ultrapure

Product Code. 15825168 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Plastic Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15825168 25 g Plastic Bottle 25g
15815168 5 g Plastic Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15825168 Supplier Thermo Scientific Supplier No. J17924.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 25389-94-0 | C18H38N4O15S | 582.575 g/mol

A selectable marker for cosmid vectors used in transfection of mammalian cells.

Mode of Action: Binds to ribosomal components and inhibits protein synthesis. Kanamycin is inactivated by an aminoglycoside phospho transferase (MW 25,000) in the periplasmic space.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Selectable marker for cosmid vectors used in transfection of mammalian cells
  • Mode of Action: Binds to ribosomal components and inhibits protein synthesis
  • Kanamycin is inactivated by aminoglycoside phospho transferase (MW 25,000) in
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25389-94-0
Molecular Formula C18H38N4O15S
Molecular Weight (g/mol) 582.575
MDL Number MFCD00070253
InChI Key OOYGSFOGFJDDHP-IZQIRFRQSA-N
Synonym Kanamycin A
PubChem CID 134129479
IUPAC Name (2 R,3 S,4 S,5 S,6 R)-2-(aminomethyl)-6-[(1 R,2 S,3 S,4 R,6 S)-4,6-diamino-3-[(2 S,3 R,4 S,5 R,6 R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O

Specifications

Type Kanamycin Sulfate
Physical Form Powder
Packaging Plastic Bottle
Quantity 25 g
Formula Weight 582.58
Chemical Name or Material Kanamycin sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.