missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kanamycin Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA175225
This item is not returnable.
View return policy
Description
Working Concentration: 0.3125 μg/mL. Sensitivity: 10 ng/mL Kanamycin produces 57% inhibition in a competitive ELISA employing Kanamycin polyclonal antibody.
Kanamycin is only found in individuals that have used or taken this drug. It is an antibiotic complex produced by Streptomyces kanamyceticus from Japanese soil. Comprises 3 components: kanamycin A, the major component, and kanamycins B and C, the minor components. [PubChem]Aminoglycosides like kanamycin irreversibly bind to specific 30S-subunit proteins and 16S rRNA. Specifically Kanamycin binds to four nucleotides of 16S rRNA and a single amino acid of protein S12. This interferes with decoding site in the vicinity of nucleotide 1400 in 16S rRNA of 30S subunit. This region interacts with the wobble base in the anticodon of tRNA. This leads to interference with the initiation complex, misreading of mRNA so incorrect amino acids are inserted into the polypeptide leading to nonfunctional or toxic peptides and the breakup of polysomes into nonfunctional monosomes. [National Center for Biotechnology Information. PubChem Compound Database; CID=6032].
Specifications
| Kanamycin | |
| Polyclonal | |
| Unconjugated | |
| C18H36N4O11; Kanamicina; Kanamycin A; Kanamycin Base; Kanamycinum | |
| Kanamycin-BTG. | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA | |
| 14.042 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Sheep | |
| Purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction