Learn More
Kanamycin monosulfate
Description
Antibiotic. Inhibits translocation during protein biosynthesis.Kanamycin monosulfate acts as an antibiotic and used to treat various infections. It is involved in the translation of prokaryotic ribosomes arises due to bacterial cell death. It is used as a selective agent in molecular biology in order to isolate transformants that are expressing a kanamycin resistance gene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Kanamycin monosulfate is the sulfate salt form of kanamycin A
- It was isolated from Streptomyces kanamyceticus
- This compound inhibits translocation during protein biosynthesis. It binds to the 30S ribosomal subunit, causing misreading of the mRNA sequence
Application
- Kanamycin monosulfate shows antibacterial activity. It is effective against gram-positive and gram-negative bacteria, as well as Mycoplasma species
- This compound is also involved in translating prokaryotic ribosomes
- In molecular biology experiments, it is commonly used to select and isolate transformants expressing the kanamycin resistance gene, and plants that have the NPT II (APH3) gene incorporated
Specifications
Specifications
| Type | Kanamycin Monosulfate |
| Physical Form | Powder |
| Color | White |
| Quantity | 10 g |
| Merck Index | 14,5281 |
| Solubility Information | Dissolve in water at 10mg/ml.Soluble in water. |
| Formula Weight | 582.58 |
| Chemical Name or Material | Kanamycin monosulfate |
Safety and Handling
- Kanamycin monosulphate
- Danger
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: Kanamycin sulphate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.