missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin monosulfate, MP Biomedicals™

Product Code. 11466412 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466412 5 g 5g
11476412 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466412 Supplier MP Biomedicals Supplier No. 0215002905
  • ONLINE EXCLUSIVE PRICE
    £92.23
    £80.75 / 5g
    Save £11.48
    12% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 25389-94-0
Molecular Formula C18H38N4O15S
Molecular Weight (g/mol) 582.575
InChI Key OOYGSFOGFJDDHP-IZQIRFRQSA-N
Synonym Kanamycin sulfate salt, Kanamycin A
PubChem CID 134129479
IUPAC Name (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O

Specifications

Type Kanamycin Monosulfate
Physical Form Powder
Color White
Quantity 5 g
Formula Weight 582.6
Chemical Name or Material Kanamycin monosulfate
compliance-icons compliance-icons
Product Identifier
  • Propidium Iodide
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.