Learn More
Kanamycin monosulfate, Cell Culture Grade
Description
Kanamycin monosulfate, is used as an aminoglycoside antibiotic. Kanamycin can be used as a selection agent in several types of isolation media.Kanamycin sulfate from Streptomyces kanamyceticus has been used in tissue culture media. Kanamycin sulfate can also be used as a selection agent for cells transformed with kanamycin B resistance gene. It is recommended for use in cell culture applications at 100mg/mL.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Kanamycin monosulfate is the sulfate salt form of kanamycin A
• It was isolated from Streptomyces kanamyceticus
• This compound inhibits translocation during protein biosynthesis. It binds to the 30S ribosomal subunit, causing misreading of the mRNA sequence
Application
• Kanamycin monosulfate shows antibacterial activity. It is effective against gram-positive and gram-negative bacteria, as well as Mycoplasma species
• This compound is also involved in translating prokaryotic ribosomes
• In molecular biology experiments, it is commonly used to select and isolate transformants expressing the kanamycin resistance gene, and plants that have the NPT II (APH3) gene incorporated
Especificaciones
Especificaciones
| Type | Kanamycin Monosulfate |
| Physical Form | Powder |
| Color | White |
| Quantity | 1 g |
| Merck Index | 14,5281 |
| Solubility Information | Soluble in water at 10mg/ml |
| Formula Weight | 582.58 |
| Grade | Cell Culture |
| Chemical Name or Material | Kanamycin monosulfate |
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.