Learn More
Kanamycin disulfate, 700μg/mg, Thermo Scientific™
£95.10
Chemical Identifiers
| CAS | 64013-70-3 |
|---|---|
| Molecular Formula | C18H40N4O19S2 |
| Molecular Weight (g/mol) | 680.647 |
| MDL Number | MFCD00143916 |
| InChI Key | OGTKIXVMLDAMNU-KNQICTBBSA-N |
| Synonym | Kanamycin A; Kanamycin acid sulfate |
| PubChem CID | 24721551 |
| IUPAC Name | (2R,3S,4S,5R,6R)-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid |
| SMILES | C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O.OS(=O)(=O)O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15428389
|
Thermo Scientific Alfa Aesar
J66856.06 |
5g |
£95.10
5g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 64013-70-3 | |
| 680.647 | |
| OGTKIXVMLDAMNU-KNQICTBBSA-N | |
| 24721551 | |
| C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O.OS(=O)(=O)O |
| C18H40N4O19S2 | |
| MFCD00143916 | |
| Kanamycin A; Kanamycin acid sulfate | |
| (2R,3S,4S,5R,6R)-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid |
Specifications
| 64013-70-3 | |
| Light and moisture sensitive | |
| MFCD00143916 | |
| Soluble in aqueous solutions | |
| C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O.OS(=O)(=O)O | |
| 680.647 | |
| 680.65 |
| Colorless | |
| C18H40N4O19S2 | |
| Kanamycin A; Kanamycin acid sulfate | |
| OGTKIXVMLDAMNU-KNQICTBBSA-N | |
| (2R,3S,4S,5R,6R)-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid | |
| 24721551 | |
| Kanamycin disulfate, 700μg/mg |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H360
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.