missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin B, TRC

Product Code. 17983625
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17983625 500 mg 500mg
17911425 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17983625 Supplier Toronto Research Chemicals Supplier No. TRCK137523500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4696-76-8
Molecular Formula C18 H37 N5 O10
Molecular Weight (g/mol) 483.51
Synonym D-Streptamine, O-3-amino-3-deoxy-α-D-glucopyranosyl-(1→6)-O-[2,6-diamino-2,6-dideoxy-α-D-glucopyranosyl-(1→4)]-2-deoxy-, Kanamycin B (7CI,8CI), O-3-Amino-3-deoxy-α-D-glucopyranosyl-(1→6)-O-[2,6-diamino-2,6-dideoxy-α-D-glucopyranosyl-(1→4)]-2-deoxy-D-streptamine, 2'-Amino-2'-deoxykanamycin, Aminodeoxykanamycin, Beckanamycin, Bekanamycin, KDM, Kanendomycin, NK 1006, Nebramycin V, Nebramycin factor 5
IUPAC Name (2R,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4-diol
SMILES NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](N)[C@@H](O)[C@@H]1O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C18H37N5O10/c19-2-6-11(26)12(27)9(23)17(30-6)32-15-4(20)1-5(21)16(14(15)29)33-18-13(28)8(22)10(25)7(3-24)31-18/h4-18,24-29H,1-3,19-23H2/t4-,5+,6+,7+,8-,9+,10+,11+,12+,13+,14-,15+,16-,17+,18+/m0/s1
Formula Weight 483.254
Percent Purity >95%
Chemical Name or Material Kanamycin B
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.