missing translation for 'onlineSavingsMsg'
Learn More

Kanamycin A Deuterated (>90%), TRC

Product Code. 30392116
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392116 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392116 Supplier Toronto Research Chemicals Supplier No. TRCK13750210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Kanamycin A Deuterated
CAS 1174286-24-8
Molecular Formula C18H36N4O11 for undeuterated
Quantity 10 mg
Synonym O-3-Amino-3-deoxy-α-D-glucopyranosyl-(16)-O-[6-amino-6-deoxy-α-D-glucopyranosyl-(14)]-2-deoxy-D-streptamine Deuterated, Cantrex Deuterated, Cristalomicina Deuterated, Enterokanacin Deuterated, Kamycin Deuterated, Kanabristol Deuterated, Kanamytrex Deuterated, Kanaqua Deuterated, Kanasig Deuterated, Kanatrol Deuterated, Kanescin Deuterated, Otokalixin Deuterated, Resistomycin Deuterated
InChI Formula InChI=1S/C18H36N4O11/c19-2-6-10(25)12(27)13(28)18(30-6)33-16-5(21)1-4(20)15(14(16)29)32-17-11(26)8(22)9(24)7(3-23)31-17/h4-18,23-29H,1-3,19-22H2/t4-,5+,6-,7-,8+,9-,10-,11-,12+,13-,14-,15+,16-,17-,18-/m1/s1
SMILES O[C@H]1[C@H](O[C@@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](CN)O2)[C@@H](N)C[C@@H](N)[C@@H]1O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O
Molecular Weight (g/mol) 484.5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.