missing translation for 'onlineSavingsMsg'
Learn More

Kanadaptin Antibody, Biotin, Novus Biologicals™

Product Code. 18364105 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364105 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364105 Supplier Novus Biologicals Supplier No. NBP306068B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Kanadaptin Polyclonal specifically detects Kanadaptin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kanadaptin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias FLJ10624, FLJ41004, HLC3, HLC-3, Human lung cancer oncogene 3 protein, kanadaptin, Kidney anion exchanger adapter protein, kidney anion exchanger adaptor protein, lung cancer oncogene 3, MGC120646, MGC120648, solute carrier family 4 (anion exchanger), member 1, adapter protein, solute carrier family 4 (anion exchanger), member 1, adaptor protein, Solute carrier family 4 anion exchanger member 1 adapter protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Kanadaptin fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22950
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.