missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KALRN Polyclonal specifically detects KALRN in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | KALRN |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | ARHGEF24, DUETFLJ16443, DUO, EC 2.7.11.1, FLJ12332, FLJ18623, HAPIP, huntingtin-associated protein interacting protein (duo), Huntingtin-associated protein-interacting protein, Kalirin, kalirin, RhoGEF kinase, Protein Duo, serine/threonine kinase with Dbl- and pleckstrin homology domains, Serine/threonine-protein kinase with Dbl- and pleckstrin homology domain, TRADFLJ18196 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human KALRN (NP_001019831.2). Peptide sequence QGDSADEKSKKGWGEDEPDEESHTPLPPPMKIFDNDPTQDEMSSLLAARQ |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?