Learn More
Invitrogen™ Kallikrein 6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588324
Description
Immunogen sequence: AASHDQDIML LRLARPAKLS ELIQPLPLER DCSANTTSCH ILGWGKTADG DFPDTIQCAY IHLVSREECE HAYPGQITQN MLCAGDEKYG KDSCQGDSGG PLVCGDHLRG LVSWGNIPCG SKEKPGVYTN VCRYTNWIQK TIQAK; Positive Samples: HT-29; Cellular Location: Cytoplasm, Microsome, Mitochondrion, Nucleus, nucleolus.
Kallikreins, including Kallikrein 6, are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. The KLK6 enzyme is regulated by steroid hormones. In tissue culture, the enzyme has been found to generate amyloidogenic fragments from the amyloid precursor protein, suggesting a potential for involvement in Alzheimer's disease.
Specifications
| Kallikrein 6 | |
| Polyclonal | |
| Unconjugated | |
| Klk6 | |
| 0610007D04Rik; AI849898; brain serine protease; BSP; Bssp; Glandular kallikrein K1; hK6; Kal; KAL-B; kallikrein 1; kallikrein 29; kallikrein 6; kallikrein 6 (neurosin); kallikrein 6 (neurosin, zyme); kallikrein 7; kallikrein related peptidase 6; kallikrein related-peptidase 6; kallikrein renal/pancreas/salivary; kallikrein-1; Kallikrein6; Kallikrein-6; Klk1; Klk1b6; Klk29; Klk6; Klk-6; Klk7; MGC9355; mGK-6; MSP; myelencephalon-specific protease; myofibril-bound protease; neurosin; Protease M; protease serine 9 (neurosin); protease, serine, 18; protease, serine, 9; PRSS18; PRSS9; renal kallikrein; Serine protease 18; serine protease 9; SP59; tissue kallikrein-6; true tissue kallikrein; zyme | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5653 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.57 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q92876 | |
| Klk6 | |
| A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human KLK6 (NP_0027651). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.