missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 5 Mouse anti-Human, Clone: KLK5/3845, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Kallikrein 5 |
|---|---|
| Clone | KLK5/3845 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
Kallikrein 5 Monoclonal specifically detects Kallikrein 5 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
| Kallikrein 5 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| 10 mM PBS with 0.05% BSA | |
| 25818 | |
| IgG2a κ | |
| Protein A or G purified |
| KLK5/3845 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.4.21, EC 3.4.21.4, kallikrein 5, Kallikrein-like protein 2, kallikrein-related peptidase 5, KLKL2, KLK-L2EC 3.4.21.-, SCTEkallikrein-5, Stratum corneum tryptic enzyme | |
| Recombinant fragment of human Kallikrein 5 protein (around aa 36-177) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title