missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 5 Antibody (KLK5/3845), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08366CL1
This item is not returnable.
View return policy
Description
Kallikrein 5 Monoclonal antibody specifically detects Kallikrein 5 in Human samples. It is validated for Immunohistochemistry (Paraffin),Protein Array
Specifications
| Kallikrein 5 | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 3.4.21, EC 3.4.21.4, kallikrein 5, Kallikrein-like protein 2, kallikrein-related peptidase 5, KLKL2, KLK-L2EC 3.4.21.-, SCTEkallikrein-5, Stratum corneum tryptic enzyme | |
| Recombinant fragment of human Kallikrein 5 protein (around aa 36-177) (exact sequence is proprietary) (Uniprot: Q9Y337) | |
| 0.1 mL | |
| Cancer | |
| 25818 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| KLK5/3845 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction