missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 5 Antibody (Genentech patent anti-KLK5), FITC, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27961F
This item is not returnable.
View return policy
Description
Kallikrein 5 Monoclonal antibody specifically detects Kallikrein 5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Kallikrein 5 | |
| Monoclonal | |
| FITC | |
| EC 3.4.21, EC 3.4.21.4, kallikrein 5, Kallikrein-like protein 2, kallikrein-related peptidase 5, KLKL2, KLK-L2EC 3.4.21.-, SCTEkallikrein-5, Stratum corneum tryptic enzyme | |
| KLK5/ Kallikrein 5 | |
| 0.1 mL | |
| Cancer | |
| 25818 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Genentech patent anti-KLK5 | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction