missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 3/PSA Antibody (KLK3/801 + KLK3/1248) - Azide and BSA Free, Novus Biologicals™

Product Code. 18361062 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361062 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361062 Supplier Novus Biologicals Supplier No. NBP311567

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Kallikrein 3/PSA Monoclonal antibody specifically detects Kallikrein 3/PSA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 3/PSA
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KLK3/801 + KLK3/1248
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Formulation 10 mM PBS
Gene Alias APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin
Host Species Mouse
Immunogen Recombinant full-length human Kallikrein 3/PSA protein
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Prostate Cancer, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 354
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1, IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.