missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kallikrein 2 Recombinant Rabbit Monoclonal Antibody (24GB3700)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557045
This item is not returnable.
View return policy
Description
Kallikrein 2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Glandular kallikreins cleave Met-Lys and Arg-Ser bonds in kininogen to release Lys-bradykinin.
Specifications
| Kallikrein 2 | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P20151 | |
| KLK2, Klk2-ps | |
| A synthesized peptide derived from human KLK2 (aa 210-261). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3700 | |
| Unconjugated | |
| KLK2 | |
| glandular kallikrein 2; glandular kallikrein-1; hGK-1; hK2; kallikrein 2, prostatic; kallikrein related peptidase 2; kallikrein-2; kallikrein-related peptidase 2; KLK2; KLK2A2; MGC12201; Tissue kallikrein 2; Tissue kallikrein-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16603, 3817 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction