missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10812-1-AP-150UL
This item is not returnable.
View return policy
Description
Glandular kallikreins cleave Met-Lys and Arg-Ser bonds in kininogen to release Lys-bradykinin.Specifications
| Kallikrein 2 | |
| Polyclonal | |
| Unconjugated | |
| KLK2 | |
| Glandular kallikrein 1, hGK 1, hK2, Kallikrein 2, kallikrein related peptidase 2, KLK2, KLK2A2, Tissue kallikrein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16603, 3817 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P20151 | |
| KLK2, Klk2-ps | |
| Kallikrein 2 Fusion Protein Ag1077 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction