missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Kallikrein 2 Antibody (JNJ-69086420), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28696-50ug
This item is not returnable.
View return policy
Description
Kallikrein 2 Monoclonal antibody specifically detects Kallikrein 2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Kallikrein 2 | |
| Monoclonal | |
| Unconjugated | |
| P20151 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| JNJ-69086420 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| EC 3.4.21, EC 3.4.21.35, glandular kallikrein 2, Glandular kallikrein-1, hGK-1, hK2, kallikrein 2, prostatic, kallikrein-2, kallikrein-related peptidase 2, KLK2, KLK2A2, MGC12201, Tissue kallikrein-2 | |
| KLK2/ Kallikrein 2 | |
| 50 μg | |
| Cancer, Signal Transduction | |
| 3817 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction