missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 2 Antibody (426723) [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30735684 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735684 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735684 Supplier Novus Biologicals Supplier No. FAB4104MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Kallikrein 2 Monoclonal antibody specifically detects Kallikrein 2 in Human samples. It is validated for Immunoprecipitation,Western Blot,Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 2
Applications Immunoprecipitation, Western Blot, Neutralization
Classification Monoclonal
Clone 426723
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.35, glandular kallikrein 2, Glandular kallikrein-1, hGK-1, hK2, kallikrein 2, prostatic, kallikrein-2, kallikrein-related peptidase 2, KLK2, KLK2A2, MGC12201, Tissue kallikrein-2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Kallikrein 2, Ile25-Pro261, Accession # P20151
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3817
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.