missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 2 Antibody (3C5), Novus Biologicals™

Product Code. 18377788 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377788 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377788 Supplier Novus Biologicals Supplier No. H00003817M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Kallikrein 2 Monoclonal antibody specifically detects Kallikrein 2 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 2
Applications ELISA, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 3C5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH05196
Gene Alias EC 3.4.21, EC 3.4.21.35, glandular kallikrein 2, Glandular kallikrein-1, hGK-1, hK2, kallikrein 2, prostatic, kallikrein-2, kallikrein-related peptidase 2, KLK2, KLK2A2, MGC12201, Tissue kallikrein-2
Host Species Mouse
Immunogen KLK2 (AAH05196, 1 a.a. ~ 262 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MWDLVLSIALSVGCTGAVPLIQSRIVGGWECEKHSQPWQVAVYSHGWAHCGGVLVHPQWVLTAAHCLKKNSQVWLGRHNLFEPEDTGQRVPVSHSFPHPLYNMSLLKHQSLRPDEDSSHDLMLLRLSEPAKITDVVKVLGLPTQEPALGTTCYASGWGSIEPEEFLRPRSLQCVSLHLLSNDMCARAYSEKVTEFMLCAGLWTGGKDTCGGDSGGPLVCNGVLQGITSWGPEPCALPEKPAVYTKVVHYRKWIKDTIAANP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3817
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.