missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 15 Antibody (MM0429-5Z12), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11749F
This item is not returnable.
View return policy
Description
Kallikrein 15 Monoclonal antibody specifically detects Kallikrein 15 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Kallikrein 15 | |
| Monoclonal | |
| FITC | |
| ACO, ACO protease, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, HSRNASPH, kallikrein 15, kallikrein-15, kallikrein-like serine protease, kallikrein-related peptidase 15, prostinogen | |
| Human recombinant Kallikrein 15 | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 55554 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0429-5Z12 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction