missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 13 Antibody (MM0428-5X29), Janelia Fluor™ 549, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Kallikrein 13 Monoclonal antibody specifically detects Kallikrein 13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
Specifications
Specifications
| Antigen | Kallikrein 13 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization |
| Classification | Monoclonal |
| Clone | MM0428-5X29 |
| Conjugate | Janelia Fluor 549 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp586J1923, EC 3.4.21, EC 3.4.21.-, kallikrein 13, Kallikrein-like protein 4, kallikrein-related peptidase 13, KLK-L4kallikrein-13, KLKL4kallikrein-like gene 4 |
| Host Species | Mouse |
| Immunogen | Human recombinant Kallikrein 13 |
| Purification Method | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?