missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kallikrein 12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14237
This item is not returnable.
View return policy
Description
PA1-4237 detects Kallikrein-12 from human samples. PA1-4237 has been successfully used in Western blot procedures. PA1-4237 immunizing peptide corresponds to amino acid residues 236-248 from human Kallikrein-12.
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Alternate splicing of this gene results in three transcript variants encoding different isoforms.
Specifications
| Kallikrein 12 | |
| Polyclonal | |
| Unconjugated | |
| KLK12 | |
| 2310008B01Rik; AI324852; kallikrein 12; kallikrein related peptidase 12; kallikrein related-peptidase 12; kallikrein-12; Kallikrein-like protein 5; kallikrein-related peptidase 12; KLK12; KLK-12; KLKL5; KLK-L5; UNQ669/PRO1303 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 43849 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9UKR0 | |
| KLK12 | |
| Synthetic Peptide: K(236) Y V D W I R M I M R N N(248). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction