missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kallikrein 12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100930
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KLK12.
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Alternate splicing of this gene results in three transcript variants encoding different isoforms.
Specifications
| Kallikrein 12 | |
| Polyclonal | |
| Unconjugated | |
| KLK12 | |
| 2310008B01Rik; AI324852; kallikrein 12; kallikrein related peptidase 12; kallikrein related-peptidase 12; kallikrein-12; Kallikrein-like protein 5; kallikrein-related peptidase 12; KLK12; KLK-12; KLKL5; KLK-L5; UNQ669/PRO1303 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 43849 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UKR0 | |
| KLK12 | |
| A synthesized peptide derived from human KLK12(Accession Q9UKR0), corresponding to amino acid residues S76-P126. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction